Search
Search

Rab3d| Gene Symbol: Rab3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031874.4 | NP_114080.2 | 3 | 499 | ||
| GenBank mRNA | AB232590.1 | - | 2 | 265 | ||
| AF263365.1 | - | 3 | 499 | |||
| AK048395.1 | - | 3 | 439 | |||
| AK078462.1 | - | 3 | 378 | |||
| AK169369.1 | - | 3 | 377 | |||
| BC010779.1 | - | 3 | 366 | 4216156 | ||
| BC020010.1 | - | 3 | 391 | 3584902 | ||
| M89777.1 | - | 2 | 308 | |||