Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL1R1 SIRNA Assays ›
          Gene Symbol:
          IL1R1
          Gene Name:
          interleukin 1 receptor type 1
          Gene Aliases:
          CD121A, D2S1473, IL-1R-alpha, IL1R, IL1RA, P80
          Chromosome Location:
          Chr.2: 102069638 - 102179874 on Build GRCh38
          UniGene ID:
          Hs.701982
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s7272

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL1R1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000877.3 NP_000868.1 6 912
          NM_001288706.1 NP_001275635.1 6 825
          NM_001320978.1 NP_001307907.1 6 825
          NM_001320980.1 NP_001307909.1 6 966
          NM_001320981.1 NP_001307910.1 6 807
          NM_001320982.1 NP_001307911.1 6 749
          NM_001320983.1 NP_001307912.1 4 600
          NM_001320984.1 NP_001307913.1 4 477
          XM_005263930.4 XP_005263987.1 6 774
          XM_005263934.4 XP_005263991.1 7 879
          XM_011511115.2 XP_011509417.1 7 871
          XM_011511116.1 XP_011509418.1 6 849
          XM_011511117.1 XP_011509419.1 6 844
          XM_011511118.2 XP_011509420.1 6 775
          XM_017003989.1 XP_016859478.1 6 972
          GenBank mRNA AK314433.1 - 6 825
          BC067506.1 - 5 646 7002107
          BC067508.1 - 5 648 7002111
          BC075062.2 - 5 628 30915370
          BC075063.2 - 5 628 30915586
          CD013912.1 - 4 469
          CD014140.1 - 4 592
          M27492.1 - 5 666
          X16896.1 - 6 892

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          cell surface receptor signaling pathway
          response to radiation
          heat acclimation
          ovulation
          regulation of inflammatory response
          interleukin-1-mediated signaling pathway
          response to interleukin-1
          response to epidermal growth factor
          cellular response to glucose stimulus
          response to transforming growth factor beta
          response to nitric oxide
          protease binding
          signal transducer activity
          transmembrane signaling receptor activity
          interleukin-1 receptor activity
          interleukin-1, Type I, activating receptor activity
          platelet-derived growth factor receptor binding
          protein binding
          ionotropic glutamate receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0