Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009077.2 | NP_033103.2 | 3 | 216 | ||
GenBank mRNA | AK009044.1 | - | 3 | 187 | ||
AK010510.1 | - | 3 | 184 | |||
AK012580.1 | - | 3 | 184 | |||
AK145649.1 | - | 3 | 217 | |||
AK151667.1 | - | 3 | 186 | |||
AK153172.1 | - | 3 | 184 | |||
AK166803.1 | - | 3 | 186 | |||
AK168288.1 | - | 3 | 184 | |||
AK169659.1 | - | 3 | 184 | |||
BC072558.1 | - | 2525 | ||||
BC081468.1 | - | 3 | 174 | 6823249 | ||
BC082290.1 | - | 3 | 170 | 6485569 | ||
BC091732.1 | - | 2 | 162 | 2655484 | ||
DQ403025.1 | - | 2 | 81 | |||
L04128.1 | - | 2 | 176 |