Search
Search

Rpn2| Gene Symbol: Rpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019642.4 | NP_062616.2 | 9 | 1238 | ||
| GenBank mRNA | AK004968.1 | - | 9 | 1239 | ||
| AK018761.2 | - | 9 | 1278 | |||
| AK087923.1 | - | 3 | 275 | |||
| AK153952.1 | - | 9 | 1099 | |||
| AK161624.1 | - | 9 | 1049 | |||
| AK166778.1 | - | 9 | 1024 | |||
| AK167864.1 | - | 9 | 1018 | |||
| AK167866.1 | - | 9 | 1018 | |||
| AK168221.1 | - | 9 | 1018 | |||
| BC010509.1 | - | 8 | 1011 | |||
| BC046806.1 | - | 9 | 1266 | 6829209 | ||
| D31717.1 | - | 8 | 1024 | |||