Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159731.1 | NP_001153203.1 | 8 | 1157 | ||
NM_009107.3 | NP_033133.1 | 9 | 1613 | |||
GenBank mRNA | AK132607.1 | - | 2 | 181 | ||
BC058401.1 | - | 9 | 1595 | 5707723 | ||
G49237.1 | - | 9 | 1345 | |||
M84819.1 | - | 9 | 1258 | |||
S62948.1 | - | 9 | 1457 | |||
S62952.1 | - | 8 | 1113 | |||
X66225.1 | - | 9 | 1345 |