Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009116.2 | NP_033142.2 | 2 | 348 | ||
GenBank mRNA | AK019971.1 | - | 315 | |||
BC137872.1 | - | 2 | 375 | 8860891 | ||
BC137874.1 | - | 2 | 375 | 8860893 | ||
CB193470.1 | - | 1 | 23 | |||
CJ179520.1 | - | 2 | 349 | |||
X52875.1 | - | 2 | 435 |