Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205321.1 | NP_001192250.1 | 11 | 1398 | ||
NM_009134.3 | NP_033160.2 | 11 | 1398 | |||
GenBank mRNA | AK135787.1 | - | 8 | 1019 | ||
AK157866.1 | - | 11 | 1398 | |||
AY538273.1 | - | 10 | 1328 |