Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SCN1B SIRNA Assays ›
          Gene Symbol:
          Scn1b
          Gene Name:
          sodium channel, voltage-gated, type I, beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 31116524 - 31126945 on Build GRCm38
          UniGene ID:
          Mm.1418
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s73369

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Scn1b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011322.2 NP_035452.1 2, 3 455
          GenBank mRNA BC009652.1 - 2, 3 455 3489368
          BC039140.1 - 3, 4 458 3672893
          U46681.1 - 2, 3 233
          U85786.1 - 2, 3 275

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: BC112922.
          This siRNA also targets the following transcripts in Rat: NM_017288, NM_001271046, NM_001271045, M91808, BC094523, and others.

          Panther Classification:

          Molecular Function -

          ion channel transporter voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of sodium ion transport
          transport
          ion transport
          sodium ion transport
          axon guidance
          positive regulation of sodium ion transport
          positive regulation of neuron projection development
          neuronal action potential propagation
          corticospinal neuron axon guidance
          regulation of ion transmembrane transport
          sodium ion transmembrane transport
          locomotion
          response to pyrethroid
          membrane depolarization
          cardiac muscle contraction
          regulation of ventricular cardiac muscle cell membrane repolarization
          regulation of atrial cardiac muscle cell membrane depolarization
          cardiac conduction
          cardiac muscle cell action potential involved in contraction
          membrane depolarization during cardiac muscle cell action potential
          membrane depolarization during Purkinje myocyte cell action potential
          regulation of heart rate by cardiac conduction
          regulation of sodium ion transmembrane transporter activity
          voltage-gated ion channel activity
          voltage-gated sodium channel activity
          sodium channel activity
          protein binding
          sodium channel regulator activity
          sodium channel inhibitor activity
          voltage-gated sodium channel activity involved in cardiac muscle cell action potential
          voltage-gated sodium channel activity involved in Purkinje myocyte action potential

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-phl7q:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0