Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009170.3 | NP_033196.1 | 1 | 595 | ||
GenBank mRNA | AK077688.1 | - | 1 | 596 | ||
BC063087.1 | - | 1 | 582 | 6516263 | ||
EU664592.1 | - | 1 | 141 | |||
X76290.1 | - | 1 | 257 |