Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009178.3 | NP_033204.2 | 8 | 700 | ||
GenBank mRNA | AB061305.1 | - | 8 | 648 | ||
AK008543.1 | - | 2 | 178 | |||
AK131874.1 | - | 2 | 178 | |||
BC011121.1 | - | 8 | 645 | 3497996 | ||
BC050773.1 | - | 8 | 700 | 6587379 | ||
D28941.1 | - | 8 | 655 | |||
X95809.1 | - | 7 | 527 |