Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6galnac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276425.1 | NP_001263354.1 | 3, 4 | 446 | ||
NM_011373.3 | NP_035503.1 | 2, 3 | 799 | |||
GenBank mRNA | AJ007310.1 | - | 3, 4 | 441 | ||
AK007601.1 | - | 3, 4 | 429 | |||
AK085730.1 | - | 3, 4 | 437 | |||
AK144884.1 | - | 3, 4 | 437 | |||
AK153036.1 | - | 3, 4 | 406 | |||
AK155956.1 | - | 3, 4 | 437 | |||
AK159712.1 | - | 2, 3 | 790 | |||
AK161559.1 | - | 3, 4 | 431 | |||
BC044734.2 | - | 2, 3 | 745 | |||
BC056451.1 | - | 2, 3 | 252 | 5685866 | ||
BC086784.1 | - | 387 | 6844232 | |||
Y15779.1 | - | 3, 4 | 423 | |||
Y15780.1 | - | 2, 3 | 435 |