Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164480.1 | NP_001157952.1 | 7 | 1452 | ||
NM_001164481.1 | NP_001157953.1 | 8 | 1706 | |||
NM_001164482.1 | NP_001157954.1 | 6 | 1391 | |||
NM_001164568.1 | NP_001158040.1 | 6 | 1421 | |||
NM_011379.4 | NP_035509.4 | 7 | 1675 | |||
GenBank mRNA | AK028151.1 | - | 6 | 662 | ||
AK137578.1 | - | 7 | 1398 | |||
AK149669.1 | - | 7 | 1398 | |||
AK156280.1 | - | 8 | 1660 | |||
AK169945.1 | - | 6 | 1337 | |||
AK170246.1 | - | 6 | 1351 | |||
AK220472.1 | - | 7 | 1588 | |||
BC054824.1 | - | 7 | 1396 | 5324326 | ||
D11374.1 | - | 6 | 1349 |