Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL15RA SIRNA Assays ›
Gene Symbol:
IL15RA
Gene Name:
interleukin 15 receptor subunit alpha
Gene Aliases:
CD215
Chromosome Location:
Chr.10: 5948897 - 5978741 on Build GRCh38
UniGene ID:
Hs.445124
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s7381
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IL15RA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243539.1
NP_001230468.1
4
816
NM_001256765.1
NP_001243694.1
5
765
NM_002189.3
NP_002180.1
4
612
NM_172200.2
NP_751950.2
3
513
NR_046362.1
2
522
XM_011519461.2
XP_011517763.1
6
1006
XM_011519462.2
XP_011517764.1
5
924
XM_011519463.2
XP_011517765.1
5
907
XM_011519464.2
XP_011517766.1
5
853
XM_011519465.1
XP_011517767.1
6
1006
XM_011519466.2
XP_011517768.1
6
1006
XM_011519467.2
XP_011517769.1
4
769
XM_011519468.1
XP_011517770.1
6
1006
XM_011519469.1
XP_011517771.1
5
907
XM_011519470.2
XP_011517772.1
3
672
XM_011519471.2
XP_011517773.1
4
737
XM_011519472.2
XP_011517774.1
3
576
XM_011519474.2
XP_011517776.1
3
559
XM_011519475.2
XP_011517777.1
2
477
XM_011519476.2
XP_011517778.1
2
467
XM_011519477.2
XP_011517779.1
2
472
XM_017016195.1
XP_016871684.1
5
853
XM_017016196.1
XP_016871685.1
4
578
XM_017016197.1
XP_016871686.1
5
593
XM_017016198.1
XP_016871687.1
4
737
XM_017016200.1
XP_016871689.1
3
1164
XM_017016201.1
XP_016871690.1
4
861
XM_017016202.1
XP_016871691.1
5
582
XM_017016203.1
XP_016871692.1
3
1164
GenBank mRNA
AF035279.1
-
4
547
AK304211.1
-
3
424
BC065506.1
-
2
514
BC074726.2
-
4
524
30915179
BC107777.1
-
4
680
6497994
BC121140.2
-
4
519
40120998
BC121141.2
-
4
524
40120999
BM546106.1
-
2
533
CR457064.1
-
3
385
CR542023.1
-
4
493
DC298692.1
-
3
518
HQ401283.1
-
5
646
JX987304.1
-
5
646
JX987305.1
-
5
646
JX987306.1
-
5
646
JX987307.1
-
5
646
JX987308.1
-
5
646
JX987309.1
-
5
646
JX987310.1
-
5
646
JX987311.1
-
5
646
U31628.1
-
4
575
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
cell proliferation
cytokine-mediated signaling pathway
signal transducer activity
cytokine receptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-5svh6:80/10.239.165.170:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0