Search
Search

Slc10a2| Gene Symbol: Slc10a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011388.2 | NP_035518.1 | 1 | 139 | ||
| GenBank mRNA | AB002693.1 | - | 1 | 139 | ||
| AY529655.1 | - | 1 | 139 | |||
| BC120703.1 | - | 1 | 120 | 40129626 | ||
| BC125290.1 | - | 1 | 120 | 40129805 | ||
| D87059.1 | - | 1 | 66 | |||