siRNA Details
Gene Symbol: Snrpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082961.1 | NP_001076430.1 | 11 | 1461 | ||
NM_001082962.1 | NP_001076431.1 | 11 | 1505 | |||
NM_013670.3 | NP_038698.1 | 10 | 1399 | |||
GenBank mRNA | AK144386.1 | - | 10 | 1355 | ||
BU708754.1 | - | 4 | 602 |
Gene Symbol: Snurf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033174.3 | NP_149409.1 | 10 | 1399 |

More Information
Panther Classification:
Molecular Function -




