siRNA Details
Gene Symbol: Sox12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011438.2 | NP_035568.1 | 1 | 1496 | ||
GenBank mRNA | CA749985.1 | - | 1 | 453 |

Gene Symbol: Sox12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011438.2 | NP_035568.1 | 1 | 1496 | ||
GenBank mRNA | CA749985.1 | - | 1 | 453 |