siRNA Details
Gene Symbol: Sox17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289464.1 | NP_001276393.1 | 4 | 2877 | ||
NM_001289465.1 | NP_001276394.1 | 4 | 3293 | |||
NM_001289466.1 | NP_001276395.1 | 4 | 2549 | |||
NM_001289467.1 | NP_001276396.1 | 3 | 2486 | |||
NM_011441.5 | NP_035571.1 | 5 | 3660 | |||
GenBank mRNA | AK004781.1 | - | 5 | 2721 | ||
D49473.1 | - | 3 | 1895 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
