Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009236.2 | NP_033262.2 | 1 | 444 | ||
GenBank mRNA | AK144281.1 | - | 1 | 445 | ||
BC006612.1 | - | 1 | 444 | 3967084 | ||
L29086.1 | - | 1 | 97 | |||
L35032.1 | - | 1 | 413 |