siRNA Details
Gene Symbol: ING1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267728.1 | NP_001254657.1 | 2 | 596 | ||
NM_005537.5 | NP_005528.4 | 2 | 1501 | |||
NM_198217.2 | NP_937860.1 | 2 | 859 | |||
NM_198218.2 | NP_937861.1 | 2 | 828 | |||
NM_198219.2 | NP_937862.1 | 2 | 1527 | |||
GenBank mRNA | AB024401.1 | - | 2 | 1090 | ||
AB024402.1 | - | 2 | 1530 | |||
AB031269.1 | - | 2 | 945 | |||
AB037594.2 | - | 2 | 596 | |||
AF001954.1 | - | 2 | 700 | |||
AF044076.1 | - | 2 | 684 | |||
AF078835.1 | - | 2 | 640 | |||
AF149721.1 | - | 2 | 1090 | |||
AF149722.1 | - | 2 | 631 | |||
AF149723.1 | - | 2 | 746 | |||
AF181849.1 | - | 2 | 1501 | |||
AF181850.1 | - | 2 | 1512 | |||
AJ310392.1 | - | 2 | 645 | |||
AK302353.1 | - | 2 | 859 | |||
BC018348.1 | - | 2 | 936 | |||
BC093942.1 | - | 2 | 692 | 7939787 | ||
BC093944.1 | - | 2 | 692 | 7939789 | ||
BQ430264.1 | - | 2 | 536 |

More Information
Other Species Targeted

Panther Classification:
