Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082552.2 | NP_001076021.1 | 7 | 1379 | ||
NM_009277.4 | NP_033303.3 | 8 | 1448 | |||
GenBank mRNA | AK141294.1 | - | 8 | 1428 | ||
AK150484.1 | - | 7 | 1343 | |||
AK152619.1 | - | 7 | 1343 | |||
AK171489.1 | - | 8 | 1413 | |||
BC010580.1 | - | 8 | 1388 | 3584654 | ||
CT010336.1 | - | 6 | 1273 | |||
L27990.1 | - | 8 | 1413 |