Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253862.1 | NP_001240791.1 | 10 | 955 | ||
NM_001253863.1 | NP_001240792.1 | 3 | 334 | |||
NM_001253864.1 | NP_001240793.1 | 3 | 334 | |||
NM_001253865.1 | NP_001240794.1 | 5 | 581 | |||
NM_011544.3 | NP_035674.2 | 10 | 1012 | |||
GenBank mRNA | AK078415.1 | - | 3 | 327 | ||
AK132002.1 | - | 3 | 319 | |||
AK133958.1 | - | 3 | 322 | |||
AK134037.1 | - | 10 | 956 | |||
AK135376.1 | - | 3 | 302 | |||
BC037097.1 | - | 5 | 569 | 5345693 | ||
CJ164494.1 | - | 3 | 336 | |||
M97635.1 | - | 10 | 983 | |||
X64840.1 | - | 10 | 812 |