Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TCF7L2 SIRNA Assays ›
          Gene Symbol:
          Tcf7l2
          Gene Name:
          transcription factor 7 like 2, T cell specific, HMG box
          Gene Aliases:
          TCF4B, TCF4E, Tcf-4, Tcf4, mTcf-4B, mTcf-4E
          Chromosome Location:
          Chr.19: 55741810 - 55933661 on Build GRCm38
          UniGene ID:
          Mm.139815
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s74838

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tcf7l2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001142918.1 NP_001136390.1 2 777
          NM_001142919.1 NP_001136391.1 2 777
          NM_001142920.1 NP_001136392.1 2 777
          NM_001142921.1 NP_001136393.1 2 777
          NM_001142922.1 NP_001136394.1 2 777
          NM_001142923.1 NP_001136395.1 2 777
          NM_009333.3 NP_033359.2 2 777
          GenBank mRNA AF107298.1 - 2 521
          AF363724.1 - 2 333
          AF363725.1 - 2 335
          AF363726.1 - 2 339
          AJ223070.1 - 2 255
          AK035059.1 - 2 605
          AK038720.1 - 2 223
          AK084488.1 - 2 318
          AK132465.1 - 2 706
          AY072035.1 - 2 318
          BC009807.1 - 2 282
          BC011397.1 - 2 756
          BC052022.1 - 2 356 5702449

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001146284, NM_001146285, NM_001198525, NM_001146286, NM_001146283, and others.
          This siRNA also targets the following transcripts in Rat: NM_001191052, XM_008760555.

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          blood vessel development
          cellular glucose homeostasis
          glycogen metabolic process
          glucose metabolic process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cell cycle arrest
          cell proliferation
          cellular response to starvation
          response to glucose
          post-embryonic development
          positive regulation of triglyceride biosynthetic process
          positive regulation of heparan sulfate proteoglycan biosynthetic process
          oligodendrocyte development
          Wnt signaling pathway
          neural tube development
          pituitary gland development
          bone mineralization
          negative regulation of BMP signaling pathway
          embryonic genitalia morphogenesis
          regulation of myelination
          positive regulation of insulin secretion
          positive regulation of protein binding
          secretory granule localization
          regulation of hormone metabolic process
          somatic stem cell population maintenance
          multicellular organism growth
          catenin import into nucleus
          regulation of gluconeogenesis by regulation of transcription from RNA polymerase II promoter
          negative regulation of fibroblast growth factor receptor signaling pathway
          odontogenesis of dentin-containing tooth
          glucose homeostasis
          positive regulation of apoptotic process
          negative regulation of sequence-specific DNA binding transcription factor activity
          maintenance of DNA repeat elements
          skin development
          canonical Wnt signaling pathway involved in positive regulation of epithelial to mesenchymal transition
          fat cell differentiation
          negative regulation of fat cell differentiation
          positive regulation of gluconeogenesis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of organ growth
          positive regulation of protein export from nucleus
          positive regulation of lipid biosynthetic process
          embryonic digestive tract morphogenesis
          embryonic hindgut morphogenesis
          myoblast fate commitment
          regulation of skeletal muscle tissue development
          regulation of smooth muscle cell proliferation
          generation of neurons
          regulation of oligodendrocyte differentiation
          positive regulation of epithelial cell proliferation
          positive regulation of protein kinase B signaling
          canonical Wnt signaling pathway
          face morphogenesis
          regulation of insulin secretion involved in cellular response to glucose stimulus
          negative regulation of canonical Wnt signaling pathway
          insulin metabolic process
          negative regulation of type B pancreatic cell apoptotic process
          negative regulation of extrinsic apoptotic signaling pathway
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II repressing transcription factor binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          beta-catenin binding
          transcription factor binding
          protein kinase binding
          nuclear hormone receptor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          gamma-catenin binding
          armadillo repeat domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0