Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TFAP2A SIRNA Assays ›
          Gene Symbol:
          Tfap2a
          Gene Name:
          transcription factor AP-2, alpha
          Gene Aliases:
          AP-2, AP2alpha, Ap-2 (a), Ap2, Ap2tf, Tcfap2a
          Chromosome Location:
          Chr.13: 40713802 - 40738238 on Build GRCm38
          UniGene ID:
          Mm.487150
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s74844

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tfap2a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001122948.2 NP_001116420.1 5 1167
          NM_001301674.1 NP_001288603.1 6 1146
          NM_011547.4 NP_035677.2 5 902
          GenBank mRNA AB221578.1 - 5 793
          AK013900.1 - 3 323
          AK053704.1 - 5 1152
          AK081627.1 - 5 912
          AK140956.1 - 5 913
          AK145772.1 - 5 857
          BC007471.1 - 5 877 3497082
          BC018226.1 - 6 1145 3983850
          U17285.1 - 5 894
          U17286.1 - 4 373
          U17287.1 - 5 1131
          U17288.1 - 4 916
          X57012.1 - 1 19
          X74216.1 - 5 863

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          kidney development
          neural tube closure
          lens morphogenesis in camera-type eye
          outflow tract morphogenesis
          keratinocyte development
          optic vesicle morphogenesis
          optic cup structural organization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          sensory perception of sound
          negative regulation of cell proliferation
          embryonic pattern specification
          embryonic body morphogenesis
          positive regulation of gene expression
          negative regulation of transcription by competitive promoter binding
          neural crest cell development
          anterior neuropore closure
          trigeminal nerve development
          oculomotor nerve formation
          forebrain neuron development
          positive regulation of cell migration
          positive regulation of bone mineralization
          response to lipopolysaccharide
          embryonic forelimb morphogenesis
          forelimb morphogenesis
          negative regulation of epidermal growth factor receptor signaling pathway
          inner ear morphogenesis
          negative regulation of apoptotic process
          negative regulation of neuron apoptotic process
          positive regulation of neuron apoptotic process
          skin development
          regulation of cell differentiation
          regulation of neuron differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          sympathetic nervous system development
          embryonic camera-type eye morphogenesis
          embryonic cranial skeleton morphogenesis
          skeletal system morphogenesis
          epidermis morphogenesis
          palate development
          lens induction in camera-type eye
          face morphogenesis
          bone morphogenesis
          eyelid development in camera-type eye
          cornea development in camera-type eye
          positive regulation of tooth mineralization
          cellular response to iron ion
          basement membrane organization
          negative regulation of reactive oxygen species metabolic process
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription coactivator activity
          RNA polymerase II transcription corepressor activity
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          protein binding
          transcription factor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0