Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IRF5 SIRNA Assays ›
Gene Symbol:
IRF5
Gene Name:
interferon regulatory factor 5
Gene Aliases:
SLEB10
Chromosome Location:
Chr.7: 128937737 - 128950042 on Build GRCh38
UniGene ID:
Hs.521181
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s7515
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IRF5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098627.3
NP_001092097.2
4
627
NM_001098629.2
NP_001092099.1
4
534
NM_001098630.2
NP_001092100.1
4
534
NM_001242452.2
NP_001229381.1
4
534
NM_032643.4
NP_116032.1
4
450
XM_005250317.3
XP_005250374.1
4
681
XM_006715974.2
XP_006716037.1
4
617
XM_011516158.2
XP_011514460.1
4
739
XM_011516159.2
XP_011514461.1
4
648
XM_011516160.1
XP_011514462.1
4
519
XM_011516161.1
XP_011514463.1
4
549
XM_011516162.1
XP_011514464.1
4
549
XM_011516163.2
XP_011514465.1
4
735
XM_011516164.1
XP_011514466.1
4
617
GenBank mRNA
AK293654.1
-
4
506
AK297083.1
-
4
493
AY504946.1
-
4
459
AY504947.1
-
4
482
AY693665.1
-
4
531
AY693666.1
-
4
461
AY693667.1
-
3
116
AY693668.1
-
4
531
AY693669.1
-
4
450
BC004139.2
-
4
450
2963923
BC004201.2
-
4
450
2963923
BU540632.1
-
3
296
BU950658.1
-
4
533
DQ277633.1
-
4
450
DQ277634.1
-
4
431
EU258895.1
-
3
287
EU258896.1
-
4
442
EU258897.1
-
4
541
EU258898.1
-
4
625
JQ950681.1
-
3
302
JQ950682.1
-
2
152
KP843527.1
-
4
510
KP843528.1
-
4
510
U51127.1
-
4
513
Back To Top
More Information
Panther Classification:
Molecular Function -
winged helix/forkhead transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
response to peptidoglycan
response to muramyl dipeptide
positive regulation of interferon-alpha production
positive regulation of interferon-beta production
positive regulation of interleukin-12 production
positive regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
defense response to virus
interferon-gamma-mediated signaling pathway
type I interferon signaling pathway
regulatory region DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0