Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011595.2 | NP_035725.1 | 2 | 623 | ||
GenBank mRNA | AB041611.2 | - | 2 | 480 | ||
AK083905.1 | - | 2 | 481 | |||
AK128943.1 | - | 2 | 482 | |||
AK147847.1 | - | 2 | 481 | |||
AY622855.1 | - | 1 | 12 | |||
BC014713.1 | - | 2 | 445 | 4239015 | ||
L19622.1 | - | 2 | 222 | |||
L27424.1 | - | 2 | 492 | |||
Z30970.1 | - | 2 | 497 |