Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009410.2 | NP_033436.1 | 16 | 2171 | ||
GenBank mRNA | AB006074.1 | - | 16 | 2172 | ||
AK029987.1 | - | 13 | 1564 | |||
AK037640.1 | - | 16 | 2072 | |||
AK164443.1 | - | 16 | 2072 | |||
AK170291.1 | - | 17 | 1978 | |||
AK171286.1 | - | 17 | 1978 | |||
BC099689.1 | - | 16 | 1986 | 30731611 |