Search
Search

Ttn| Gene Symbol: Ttn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011652.3 | NP_035782.3 | 8 | 1589 | ||
| NM_028004.2 | NP_082280.2 | 8 | 1589 | |||
| GenBank mRNA | AK052859.1 | - | 8 | 1589 | ||
| AK084709.1 | - | 8 | 1589 | |||
| AK084718.1 | - | 8 | 1585 | |||
| AK085832.1 | - | 8 | 1589 | |||