Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011669.3 | NP_035799.1 | 5, 6 | 946 | ||
GenBank mRNA | AF022792.1 | - | 2, 3 | 215 | ||
AF441835.1 | - | 4, 5 | 607 | |||
AK006739.1 | - | 5, 6 | 996 | |||
AK035629.1 | - | 5, 6 | 787 | |||
AK167153.1 | - | 5, 6 | 786 | |||
BC049274.1 | - | 3, 4 | 613 | 4500832 | ||
BC055398.1 | - | 1, 2 | 38 | |||
BC068136.1 | - | 5, 6 | 946 | 30363916 |