Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083319.1 | NP_001076788.1 | 15, 16 | 1945 | ||
NM_013699.2 | NP_038727.1 | 14, 15 | 1837 | |||
GenBank mRNA | AB221581.1 | - | 13, 14 | 1425 | ||
AK079815.1 | - | 7, 8 | 672 | |||
AK140942.1 | - | 9, 10 | 842 | |||
AY182062.1 | - | 14, 15 | 1534 | |||
BC034749.1 | - | 13, 14 | 2164 | |||
BC139828.1 | - | 13, 14 | 1454 | 40142981 | ||
BC141082.1 | - | 13, 14 | 1516 | 9055694 | ||
U20086.1 | - | 13, 14 | 1428 |