Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NR1H2 SIRNA Assays ›
          Gene Symbol:
          Nr1h2
          Gene Name:
          nuclear receptor subfamily 1, group H, member 2
          Gene Aliases:
          AI194859, LXR, LXRB, LXRbeta, NER1, OR-1, RIP15, UR, Unr, Unr2
          Chromosome Location:
          Chr.7: 44549616 - 44553969 on Build GRCm38
          UniGene ID:
          Mm.968
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s75786

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nr1h2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001285517.1 NP_001272446.1 8, 9 1298
          NM_001285518.1 NP_001272447.1 8, 9 1292
          NM_001285519.1 NP_001272448.1 8, 9 1289
          NM_009473.3 NP_033499.1 8, 9 1305
          GenBank mRNA AK077620.1 - 8, 9 1271
          AK134489.1 - 8, 9 1255
          AK156777.1 - 8, 9 1273
          AK177488.1 - 1, 2 56
          AK194055.1 - 1, 2 56
          BC066025.1 - 8, 9 1256 6829527
          BC106148.1 - 7, 8 1025
          EU869275.2 - 5, 6 1284
          U09419.1 - 7, 8 1123

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of triglyceride biosynthetic process
          positive regulation of cholesterol efflux
          positive regulation of lipid storage
          negative regulation of cholesterol storage
          intracellular receptor signaling pathway
          positive regulation of cellular protein metabolic process
          negative regulation of lipid transport
          positive regulation of cholesterol transport
          cholesterol homeostasis
          steroid hormone mediated signaling pathway
          cellular lipid metabolic process
          positive regulation of fatty acid biosynthetic process
          negative regulation of proteolysis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          retinoic acid receptor signaling pathway
          negative regulation of pinocytosis
          positive regulation of lipoprotein lipase activity
          lipid homeostasis
          positive regulation of high-density lipoprotein particle assembly
          positive regulation of pancreatic juice secretion
          positive regulation of secretion of lysosomal enzymes
          regulation of cholesterol homeostasis
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor activity, sequence-specific transcription regulatory region DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          zinc ion binding
          apolipoprotein A-I receptor binding
          sequence-specific DNA binding
          metal ion binding
          retinoid X receptor binding
          ATPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0