Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011712.2 | NP_035842.1 | 3 | 494 | ||
GenBank mRNA | AK002214.1 | - | 3 | 448 | ||
AK017717.1 | - | 3 | 454 | |||
AK213578.1 | - | 1 | 101 | |||
AY459189.1 | - | 3 | 493 | |||
BC007478.1 | - | 3 | 391 | 3490650 | ||
BC020289.1 | - | 3 | 419 | |||
BU511227.1 | - | 3 | 525 | |||
U92454.1 | - | 1 | 389 |