Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291747.1 | NP_001278676.1 | 3 | 171 | ||
NM_001291748.1 | NP_001278677.1 | 3 | 179 | |||
NM_011725.4 | NP_035855.1 | 3 | 219 | |||
GenBank mRNA | AA623498.1 | - | 3 | 133 | ||
AK012549.1 | - | 3 | 210 | |||
AK035025.1 | - | 3 | 181 | |||
AK035341.1 | - | 3 | 188 | |||
AK088930.1 | - | 3 | 123 | |||
BC005560.1 | - | 3 | 206 | 3965533 | ||
X07967.1 | - | 3 | 154 |
Gene Symbol: 3830403N18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014417.1 | - | 2 | 199 |