Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009536.4 | NP_033562.3 | 4 | 662 | ||
GenBank mRNA | AF483478.1 | - | 4 | 521 | ||
AF483479.1 | - | 4 | 521 | |||
AK011612.1 | - | 1 | 31 | |||
AK075995.1 | - | 4 | 611 | |||
AK135831.1 | - | 4 | 590 | |||
AK135867.1 | - | 4 | 590 | |||
AK169812.1 | - | 4 | 617 | |||
AK179917.1 | - | 2 | 284 | |||
AK183177.1 | - | 2 | 267 | |||
AK188179.1 | - | 2 | 287 | |||
AK188405.1 | - | 2 | 264 | |||
AK188896.1 | - | 1 | 159 | |||
AK190793.1 | - | 2 | 274 | |||
AK191008.1 | - | 2 | 252 | |||
AK192790.1 | - | 2 | 266 | |||
AK195324.1 | - | 2 | 267 | |||
AK195513.1 | - | 2 | 266 | |||
AK201131.1 | - | 1 | 160 | |||
AK207068.1 | - | 2 | 227 | |||
AK218242.1 | - | 2 | 240 | |||
BC058686.1 | - | 4 | 621 | 6411635 | ||
D87663.1 | - | 4 | 549 | |||
Z19599.1 | - | 4 | 604 |