Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009561.2 | NP_033587.2 | 7 | 2232 | ||
GenBank mRNA | AK148158.1 | - | 7 | 2050 | ||
AK149960.1 | - | 7 | 2027 | |||
BC006618.1 | - | 7 | 2271 | 3967527 | ||
BY420227.1 | - | 1 | 278 |