Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JAK1 SIRNA Assays ›
          Gene Symbol:
          JAK1
          Gene Name:
          Janus kinase 1
          Gene Aliases:
          JAK1A, JAK1B, JTK3
          Chromosome Location:
          Chr.1: 64833223 - 65067746 on Build GRCh38
          UniGene ID:
          Hs.207538
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s7647
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: JAK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320923.1 NP_001307852.1 22 3077
          NM_001321852.1 NP_001308781.1 21 3080
          NM_001321853.1 NP_001308782.1 23 3339
          NM_001321854.1 NP_001308783.1 22 3255
          NM_001321855.1 NP_001308784.1 22 3258
          NM_001321856.1 NP_001308785.1 21 2993
          NM_001321857.1 NP_001308786.1 21 3171
          NM_002227.3 NP_002218.2 21 3174
          XM_017001212.1 XP_016856701.1 21 3018
          GenBank mRNA AB209057.1 - 20 2930
          AB219242.1 - 19 2821
          AK314257.1 - 22 3078
          BC132729.1 - 20 2892 40146751
          BX647675.1 - 7 778
          HQ628631.1 - 19 2854
          M64174.1 - 20 2899

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00233820_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein phosphorylation
          transmembrane receptor protein tyrosine kinase signaling pathway
          cell migration
          cell differentiation
          intracellular signal transduction
          peptidyl-tyrosine autophosphorylation
          interleukin-2-mediated signaling pathway
          regulation of cell proliferation
          positive regulation of GTPase activity
          innate immune response
          response to antibiotic
          interferon-gamma-mediated signaling pathway
          regulation of interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          regulation of type I interferon-mediated signaling pathway
          positive regulation of sprouting angiogenesis
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          Ras guanyl-nucleotide exchange factor activity
          receptor binding
          growth hormone receptor binding
          protein binding
          ATP binding
          protein phosphatase binding
          ubiquitin protein ligase binding
          CCR5 chemokine receptor binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-slwzk:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0