Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JUN SIRNA Assays ›
          Gene Symbol:
          JUN
          Gene Name:
          Jun proto-oncogene, AP-1 transcription factor subunit
          Gene Aliases:
          AP-1, AP1, c-Jun
          Chromosome Location:
          Chr.1: 58780791 - 58784113 on Build GRCh38
          UniGene ID:
          Hs.696684
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s7659

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: JUN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002228.3 NP_002219.1 1 1851
          GenBank mRNA BC002646.2 - 1 1754
          BC006175.1 - 1 1244 4053956
          BC009874.2 - 1 510
          BC068522.1 - 1 1784 5267423
          BT019759.1 - 1 808
          CR541724.1 - 1 808
          CR541752.1 - 1 808

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          microglial cell activation
          release of cytochrome c from mitochondria
          liver development
          positive regulation of endothelial cell proliferation
          outflow tract morphogenesis
          transcription from RNA polymerase II promoter
          transforming growth factor beta receptor signaling pathway
          SMAD protein import into nucleus
          Ras protein signal transduction
          aging
          learning
          circadian rhythm
          negative regulation of cell proliferation
          response to radiation
          response to mechanical stimulus
          positive regulation of epithelial cell migration
          regulation of cell death
          monocyte differentiation
          axon regeneration
          negative regulation of protein autophosphorylation
          response to lipopolysaccharide
          cellular response to hormone stimulus
          response to cytokine
          leading edge cell differentiation
          response to muscle stretch
          Fc-epsilon receptor signaling pathway
          regulation of cell proliferation
          response to drug
          response to hydrogen peroxide
          negative regulation of DNA binding
          negative regulation of neuron apoptotic process
          positive regulation of neuron apoptotic process
          positive regulation of GTPase activity
          negative regulation by host of viral transcription
          positive regulation by host of viral transcription
          positive regulation of cell differentiation
          positive regulation of monocyte differentiation
          positive regulation of DNA replication
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fibroblast proliferation
          positive regulation of smooth muscle cell proliferation
          regulation of sequence-specific DNA binding transcription factor activity
          cellular response to potassium ion starvation
          response to cAMP
          regulation of cell cycle
          membrane depolarization
          SMAD protein signal transduction
          eyelid development in camera-type eye
          positive regulation of ERK1 and ERK2 cascade
          cellular response to calcium ion
          positive regulation of pri-miRNA transcription from RNA polymerase II promoter
          negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          positive regulation of DNA-templated transcription, initiation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II activating transcription factor binding
          transcriptional activator activity, RNA polymerase II transcription factor binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          transcription coactivator activity
          GTPase activator activity
          protein binding
          transcription factor binding
          enzyme binding
          cAMP response element binding
          identical protein binding
          protein homodimerization activity
          transcription regulatory region DNA binding
          poly(A) RNA binding
          protein heterodimerization activity
          R-SMAD binding
          HMG box domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0