Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011845.2 | NP_035975.1 | 6 | 1181 | ||
GenBank mRNA | AF196480.1 | - | 6 | 1181 | ||
AK039479.1 | - | 2 | 789 | |||
Y18881.1 | - | 7 | 1536 |