Search
Search

Nr0b2| Gene Symbol: Nr0b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011850.2 | NP_035980.1 | 1 | 543 | ||
| GenBank mRNA | AK004930.1 | - | 1 | 553 | ||
| AY360323.1 | - | 1 | 508 | |||
| AY360324.1 | - | 1 | 508 | |||
| BC019540.1 | - | 1 | 543 | 4193866 | ||