Search
Search

Lypla2| Gene Symbol: Lypla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011942.1 | NP_036072.1 | 6, 7 | 407 | ||
| GenBank mRNA | AB009653.1 | - | 6, 7 | 407 | ||
| AK003689.1 | - | 6, 7 | 374 | |||
| AK075590.1 | - | 6, 7 | 399 | |||
| AK089112.1 | - | 6, 7 | 399 | |||
| AK167478.1 | - | 6, 7 | 394 | |||
| BC068120.1 | - | 6, 7 | 433 | 5714650 | ||