siRNA Details
Gene Symbol: Prnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126338.2 | NP_001119810.1 | 2 | 273 | ||
NM_001278257.1 | NP_001265186.1 | 2 | 273 | |||
NM_001278520.1 | NP_001265449.1 | 2 | 252 | |||
NM_023043.3 | NP_075530.1 | 2 | 273 | |||
GenBank mRNA | AF165165.1 | - | 2 | 252 | ||
AF165166.1 | - | 2 | 256 | |||
AF192384.1 | - | 2 | 272 | |||
AF192385.1 | - | 2 | 272 | |||
AK043023.1 | - | 2 | 274 | |||
AK161043.1 | - | 2 | 274 | |||
BC025140.1 | - | 2 | 249 | 3600731 | ||
DQ832281.1 | - | 2 | 274 | |||
GQ245668.1 | - | 1 | 211 |
Gene Symbol: Prn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278258.1 | NP_001265187.1 | 4 | 495 | ||
NM_001278259.1 | NP_001265188.1 | 4 | 495 |

More Information
Other Species Targeted

