Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164089.1 | NP_001157561.1 | 3 | 398 | ||
NM_012003.2 | NP_036133.1 | 3 | 383 | |||
GenBank mRNA | AF071316.1 | - | 3 | 385 | ||
AK010417.1 | - | 3 | 393 | |||
AK010726.1 | - | 3 | 368 | |||
AK013155.1 | - | 3 | 351 | |||
AK019122.1 | - | 3 | 375 | |||
BC003724.1 | - | 2 | 264 | 3593979 | ||
BG085353.2 | - | 3 | 383 | |||
CJ165979.1 | - | 3 | 379 |