Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx3y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012008.2 | NP_036138.1 | 13 | 1431 | ||
GenBank mRNA | AJ007376.1 | - | 13 | 1398 | ||
AK154382.1 | - | 13 | 1391 | |||
BC021453.1 | - | 13 | 1396 | 5043470 | ||
BC051046.1 | - | 13 | 1420 | |||
L25337.1 | - | 5 | 496 |