Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Deb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026794.2 | NP_081070.1 | 2 | 120 | ||
GenBank mRNA | AK003863.1 | - | 2 | 128 | ||
AK049702.1 | - | 2 | 162 | |||
AK078209.1 | - | 1 | 232 | |||
AK085962.1 | - | 1 | 211 | |||
BC027548.1 | - | 2 | 120 | 1532650 |