Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2s3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012010.3 | NP_036140.1 | 10 | 1105 | ||
GenBank mRNA | AJ006587.1 | - | 10 | 1095 | ||
AK010819.1 | - | 7 | 783 | |||
AK128949.1 | - | 10 | 1104 | |||
AK145741.1 | - | 1 | 32 | |||
AK146900.1 | - | 10 | 1146 | |||
AK150095.1 | - | 10 | 1112 | |||
AK151579.1 | - | 6 | 661 | |||
AK153568.1 | - | 6 | 661 | |||
AK165866.1 | - | 10 | 1107 | |||
AK184925.1 | - | 1 | 90 | |||
BC036993.1 | - | 10 | 1103 | |||
BC063755.1 | - | 10 | 1105 | 6513251 | ||
BC139234.1 | - | 10 | 1105 | 8862256 | ||
BC139235.1 | - | 10 | 1105 | 8862257 |