Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RACGAP1 SIRNA Assays ›
          Gene Symbol:
          Racgap1
          Gene Name:
          Rac GTPase-activating protein 1
          Gene Aliases:
          AI227039, AI327394, Band25, GTPase, MgcRacGAP, gtl11, mKIAA1478
          Chromosome Location:
          Chr.15: 99620496 - 99651656 on Build GRCm38
          UniGene ID:
          Mm.273804
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s77408

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Racgap1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001253808.1 NP_001240737.1 15 1824
          NM_001253809.1 NP_001240738.1 15 1820
          NM_012025.7 NP_036155.1 16 1955
          GenBank mRNA AB030252.1 - 15 1804
          AF079974.1 - 15 1765
          AF212320.1 - 15 1791
          AF212321.1 - 16 1922
          AK144608.1 - 16 1923
          AK144745.1 - 15 1785
          AK154929.1 - 16 1943
          AK165897.1 - 15 1777
          AK166084.1 - 16 1913
          AK166094.1 - 15 1782
          AK168020.1 - 15 1781
          AK168025.1 - 15 1781
          AK168170.1 - 15 1781
          AK168679.1 - 15 1765
          AK178051.1 - 1 101
          AK180222.1 - 1 101
          AK184181.1 - 1 101
          AK187725.1 - 1 101
          AK189581.1 - 1 101
          AK205410.1 - 1 101
          AK205581.1 - 1 101
          AK205851.1 - 1 101
          AK205852.1 - 1 101
          AK206532.1 - 1 101
          AK206759.1 - 1 101
          AK207312.1 - 1 101
          AK207912.1 - 1 101
          AK208743.1 - 1 101
          AK209111.1 - 1 101
          AK209150.1 - 1 101
          AK210583.1 - 1 101
          AK211084.1 - 1 101
          AK211478.1 - 1 101
          AK212909.1 - 1 101
          AK213168.1 - 1 101
          AK213713.1 - 1 101
          AK213865.1 - 1 101
          AK214213.1 - 1 101
          AK214303.1 - 1 101
          AK214322.1 - 1 101
          AK214778.1 - 1 101
          AK215060.1 - 1 101
          AK216447.1 - 1 101
          AK216495.1 - 1 101
          AK216776.1 - 1 101
          AK217226.1 - 1 101
          AK217529.1 - 1 101
          AK217704.1 - 1 101
          AK217810.1 - 1 101
          AK218024.1 - 1 101
          AK218689.1 - 1 101
          AK218767.1 - 1 101
          AK218990.1 - 1 101
          AK219849.1 - 1 101
          BC010715.1 - 15 1770 3602242

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          actomyosin contractile ring assembly
          transport
          ion transport
          cell cycle
          signal transduction
          multicellular organism development
          spermatogenesis
          sulfate transport
          embryo development
          cell differentiation
          positive regulation of cytokinesis
          intracellular signal transduction
          positive regulation of GTPase activity
          mitotic spindle midzone assembly
          cell division
          regulation of attachment of spindle microtubules to kinetochore
          mitotic spindle assembly
          GTPase activator activity
          phosphatidylinositol-3,4,5-trisphosphate binding
          microtubule binding
          lipid binding
          protein kinase binding
          alpha-tubulin binding
          gamma-tubulin binding
          metal ion binding
          beta-tubulin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8kj9j:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0