Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: St6galnac5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012028.4 | NP_036158.3 | 4 | 935 | ||
GenBank mRNA | AB028840.1 | - | 917 | |||
AB030836.1 | - | 4 | 938 | |||
AK034387.1 | - | 939 | ||||
AK038434.1 | - | 4 | 939 | |||
AK042683.1 | - | 4 | 975 | |||
BC055737.1 | - | 4 | 935 | 6415191 |