Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC9A3R1 SIRNA Assays ›
          Gene Symbol:
          Slc9a3r1
          Gene Name:
          solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 1
          Gene Aliases:
          EBP-50, NHE-RF, NHERF-1, NHERF1
          Chromosome Location:
          Chr.11: 115163312 - 115181183 on Build GRCm38
          UniGene ID:
          Mm.27842
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s77422

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Slc9a3r1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012030.2 NP_036160.1 5, 6 1060
          GenBank mRNA AK085210.1 - 5, 6 1062
          AK153558.1 - 5, 6 1060
          AK168894.1 - 5, 6 1061
          AK177064.1 - 3, 4 241
          AK205456.1 - 3, 4 240
          AK209994.1 - 3, 4 160
          AK216315.1 - 3, 4 157
          BC085141.1 - 5, 6 1045 6413140

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          renal sodium ion transport
          adenylate cyclase-activating dopamine receptor signaling pathway
          sensory perception of sound
          negative regulation of cell proliferation
          negative regulation of platelet-derived growth factor receptor signaling pathway
          negative regulation of sodium ion transport
          negative regulation of phosphatidylinositol 3-kinase signaling
          Wnt signaling pathway
          cAMP-mediated signaling
          microvillus assembly
          actin cytoskeleton organization
          negative regulation of cell migration
          cellular phosphate ion homeostasis
          negative regulation of sodium:proton antiporter activity
          bile acid secretion
          cellular protein localization
          glutathione transport
          regulation of excretion
          regulation of protein kinase activity
          negative regulation of protein kinase B signaling
          auditory receptor cell stereocilium organization
          phospholipase C-activating dopamine receptor signaling pathway
          renal absorption
          negative regulation of ERK1 and ERK2 cascade
          renal phosphate ion absorption
          negative regulation of cell motility
          positive regulation of intrinsic apoptotic signaling pathway
          receptor binding
          protein binding
          beta-catenin binding
          chloride channel regulator activity
          phosphatase binding
          protein domain specific binding
          PDZ domain binding
          beta-2 adrenergic receptor binding
          protein complex binding
          protein complex scaffold
          protein self-association
          dopamine receptor binding
          growth factor receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-57bf8:80/100.66.79.204:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0