Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021012.4 | NP_066292.2 | 3 | 1713 | ||
XM_005256625.4 | XP_005256682.1 | 3 | 1486 | |||
XM_011523831.2 | XP_011522133.1 | 3 | 1486 | |||
GenBank mRNA | AB182123.1 | - | 1 | 1030 | ||
BC027982.1 | - | 3 | 1724 | 3873918 | ||
L36069.1 | - | 1 | 1059 |
Gene Symbol: KCNJ18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194958.2 | NP_001181887.2 | 3 | 1378 | ||
XM_005276919.3 | XP_005276976.1 | 2 | 1793 |