Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twistnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172253.2 | NP_758457.1 | 4 | 656 | ||
GenBank mRNA | AK008677.1 | - | 4 | 638 | ||
AK010829.1 | - | 4 | 657 | |||
AK016638.1 | - | 4 | 653 | |||
AK017810.1 | - | 4 | 641 | |||
AK153090.1 | - | 646 | ||||
AK160682.1 | - | 4 | 644 | |||
BC082580.1 | - | 628 | ||||
BC139162.1 | - | 4 | 644 | 8862184 | ||
BC139163.1 | - | 4 | 644 | 8862185 |