Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013914.2 | NP_038942.1 | 3 | 841 | ||
GenBank mRNA | AF133714.1 | - | 3 | 763 | ||
AF195655.1 | - | 3 | 763 | |||
AK042234.1 | - | 3 | 841 | |||
AK089285.1 | - | 3 | 830 | |||
AK153633.1 | - | 3 | 883 | |||
AK154585.1 | - | 2 | 519 | |||
BC100724.1 | - | 3 | 790 | 40045286 | ||
BC100726.1 | - | 3 | 790 | 40045284 | ||
BC100727.1 | - | 3 | 790 | 40045283 |